Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34342931-34343435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOP10
Entrez Gene ID
55505
Gene Name
NOP10 ribonucleoprotein
Gene Aliases
DKCB1, NOLA3, NOP10P
UniGene
Hs.14317
Gene Chromosome Location
Chr.15: 34341716-34343161 on Build GRCh38
Transcripts
NM_018648.3 NM_001284292.1 NM_001284293.1 NM_175741.2

Gene Symbol
NUTM1
Entrez Gene ID
256646
Gene Name
NUT midline carcinoma family member 1
Gene Aliases
C15orf55, FAM22H, NUT
UniGene
Hs.525769
Gene Chromosome Location
Chr.15: 34343315-34357737 on Build GRCh38
Transcripts
NM_018648.3 NM_001284292.1 NM_001284293.1 NM_175741.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1608281 c.13T>C p.Y5H Chr.15: 34343061-34343061 on Build GRCh38
1608282 c.6T>C p.F2F Chr.15: 34343068-34343068 on Build GRCh38
4768293 c.7C>T p.L3F Chr.15: 34343067-34343067 on Build GRCh38
5454334 c.13T>A p.Y5N Chr.15: 34343061-34343061 on Build GRCh38
3754273 c.34G>C p.D12H Chr.15: 34343040-34343040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535399943 Chr.15: 34343289-34343289 on Build GRCh38
rs568277643 Chr.15: 34343249-34343249 on Build GRCh38
rs956970210 Chr.15: 34343270-34343270 on Build GRCh38
rs1018958255 Chr.15: 34342981-34342981 on Build GRCh38
rs201920207 Chr.15: 34343092-34343092 on Build GRCh38
rs2169480 Chr.15: 34343168-34343168 on Build GRCh38
rs989314968 Chr.15: 34343276-34343276 on Build GRCh38
rs911775489 Chr.15: 34343349-34343349 on Build GRCh38
rs202190748 Chr.15: 34343011-34343011 on Build GRCh38
rs752872403 Chr.15: 34343003-34343004 on Build GRCh38
rs773321163 Chr.15: 34342991-34342991 on Build GRCh38
rs965754546 Chr.15: 34343390-34343390 on Build GRCh38
rs771140244 Chr.15: 34342994-34342994 on Build GRCh38
rs969511477 Chr.15: 34343292-34343292 on Build GRCh38
rs561915236 Chr.15: 34343147-34343147 on Build GRCh38
rs146261631 Chr.15: 34343040-34343040 on Build GRCh38
rs1003290814 Chr.15: 34343345-34343345 on Build GRCh38
rs373678744 Chr.15: 34343047-34343047 on Build GRCh38
rs1023617912 Chr.15: 34343214-34343214 on Build GRCh38
rs761222362 Chr.15: 34343023-34343023 on Build GRCh38
rs200128951 Chr.15: 34342993-34342993 on Build GRCh38
rs564679077 Chr.15: 34343091-34343091 on Build GRCh38
rs371981891 Chr.15: 34342985-34342985 on Build GRCh38
rs149606664 Chr.15: 34343179-34343179 on Build GRCh38
rs781535997 Chr.15: 34342975-34342976 on Build GRCh38
rs775092898 Chr.15: 34343130-34343130 on Build GRCh38
rs190014186 Chr.15: 34343330-34343330 on Build GRCh38
rs756047969 Chr.15: 34343050-34343050 on Build GRCh38
rs1023591694 Chr.15: 34343104-34343104 on Build GRCh38
rs35359500 Chr.15: 34343200-34343201 on Build GRCh38
rs560253884 Chr.15: 34343022-34343022 on Build GRCh38
rs989282458 Chr.15: 34343144-34343144 on Build GRCh38
rs931846866 Chr.15: 34343165-34343165 on Build GRCh38
rs186487749 Chr.15: 34343198-34343198 on Build GRCh38
rs76537972 Chr.15: 34343190-34343190 on Build GRCh38
rs1023789981 Chr.15: 34343077-34343077 on Build GRCh38
rs769790691 Chr.15: 34342988-34342988 on Build GRCh38
rs777783319 Chr.15: 34343053-34343053 on Build GRCh38
rs76029512 Chr.15: 34343192-34343192 on Build GRCh38
rs756134994 Chr.15: 34343068-34343068 on Build GRCh38
rs550497412 Chr.15: 34343138-34343138 on Build GRCh38
rs998890265 Chr.15: 34343207-34343207 on Build GRCh38
rs764894891 Chr.15: 34343089-34343089 on Build GRCh38
rs373689981 Chr.15: 34343087-34343087 on Build GRCh38
rs757349587 Chr.15: 34343124-34343124 on Build GRCh38
rs182278761 Chr.15: 34343398-34343398 on Build GRCh38
rs750840822 Chr.15: 34343179-34343180 on Build GRCh38
rs769650945 Chr.15: 34343203-34343203 on Build GRCh38
rs752462785 Chr.15: 34343132-34343132 on Build GRCh38
rs986028549 Chr.15: 34343346-34343346 on Build GRCh38
rs866026885 Chr.15: 34343159-34343159 on Build GRCh38
rs201270793 Chr.15: 34343103-34343103 on Build GRCh38
rs75061777 Chr.15: 34343396-34343396 on Build GRCh38
rs759897365 Chr.15: 34343003-34343003 on Build GRCh38
rs746040045 Chr.15: 34343083-34343083 on Build GRCh38
rs751282990 Chr.15: 34343096-34343096 on Build GRCh38
rs780995453 Chr.15: 34343048-34343048 on Build GRCh38
rs897542357 Chr.15: 34343184-34343184 on Build GRCh38
rs554420284 Chr.15: 34343123-34343123 on Build GRCh38
rs557513847 Chr.15: 34343389-34343389 on Build GRCh38
rs139502071 Chr.15: 34343182-34343182 on Build GRCh38
rs752630312 Chr.15: 34343049-34343049 on Build GRCh38
rs947285661 Chr.15: 34343054-34343054 on Build GRCh38
rs370148546 Chr.15: 34343106-34343106 on Build GRCh38
rs1030909987 Chr.15: 34343209-34343209 on Build GRCh38
rs749945545 Chr.15: 34343034-34343034 on Build GRCh38
rs749358405 Chr.15: 34343075-34343075 on Build GRCh38
rs911548653 Chr.15: 34343173-34343173 on Build GRCh38
rs556453929 Chr.15: 34343230-34343230 on Build GRCh38
rs2169479 Chr.15: 34342999-34342999 on Build GRCh38
rs368189392 Chr.15: 34342977-34342977 on Build GRCh38
rs779135425 Chr.15: 34343079-34343079 on Build GRCh38
rs972107542 Chr.15: 34343393-34343393 on Build GRCh38
rs777218307 Chr.15: 34343094-34343094 on Build GRCh38
rs533257213 Chr.15: 34343189-34343189 on Build GRCh38
rs753733230 Chr.15: 34343120-34343120 on Build GRCh38
rs764689942 Chr.15: 34343033-34343033 on Build GRCh38
rs112556317 Chr.15: 34343111-34343111 on Build GRCh38
rs370894572 Chr.15: 34343043-34343043 on Build GRCh38
rs762560152 Chr.15: 34343095-34343095 on Build GRCh38
rs181783900 Chr.15: 34343078-34343078 on Build GRCh38
rs772371219 Chr.15: 34343086-34343086 on Build GRCh38
rs762749739 Chr.15: 34343036-34343036 on Build GRCh38
rs1048581115 Chr.15: 34343402-34343402 on Build GRCh38

View Full Product Details