Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3589310-3589818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
UniGene
Hs.143681
Gene Chromosome Location
Chr.16: 3581181-3611598 on Build GRCh38
Transcripts
NM_032444.2 XM_011522715.2 XM_017023775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2919683 c.4031C>T p.S1344F Chr.16: 3589607-3589607 on Build GRCh38
4129050 c.3943A>G p.T1315A Chr.16: 3589695-3589695 on Build GRCh38
7257951 c.4045G>A p.G1349R Chr.16: 3589593-3589593 on Build GRCh38
6144618 c.3896G>T p.R1299M Chr.16: 3589742-3589742 on Build GRCh38
5505891 c.3873G>A p.T1291T Chr.16: 3589765-3589765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556682517 Chr.16: 3589712-3589712 on Build GRCh38
rs749433774 Chr.16: 3589779-3589779 on Build GRCh38
rs746416289 Chr.16: 3589428-3589428 on Build GRCh38
rs770241338 Chr.16: 3589508-3589508 on Build GRCh38
rs768415277 Chr.16: 3589582-3589582 on Build GRCh38
rs146837290 Chr.16: 3589727-3589727 on Build GRCh38
rs755683926 Chr.16: 3589534-3589534 on Build GRCh38
rs79174372 Chr.16: 3589523-3589523 on Build GRCh38
rs768464573 Chr.16: 3589447-3589447 on Build GRCh38
rs753327971 Chr.16: 3589669-3589669 on Build GRCh38
rs553317304 Chr.16: 3589487-3589487 on Build GRCh38
rs375108717 Chr.16: 3589541-3589541 on Build GRCh38
rs748691881 Chr.16: 3589397-3589397 on Build GRCh38
rs776332160 Chr.16: 3589337-3589337 on Build GRCh38
rs17136464 Chr.16: 3589539-3589539 on Build GRCh38
rs756477852 Chr.16: 3589458-3589460 on Build GRCh38
rs775530786 Chr.16: 3589502-3589502 on Build GRCh38
rs763914156 Chr.16: 3589742-3589743 on Build GRCh38
rs754996241 Chr.16: 3589797-3589797 on Build GRCh38
rs375172024 Chr.16: 3589688-3589688 on Build GRCh38
rs747230215 Chr.16: 3589617-3589617 on Build GRCh38
rs770020797 Chr.16: 3589710-3589710 on Build GRCh38
rs151144102 Chr.16: 3589592-3589592 on Build GRCh38
rs776760390 Chr.16: 3589619-3589619 on Build GRCh38
rs747719226 Chr.16: 3589459-3589459 on Build GRCh38
rs140051968 Chr.16: 3589614-3589614 on Build GRCh38
rs142205392 Chr.16: 3589581-3589581 on Build GRCh38
rs754761366 Chr.16: 3589766-3589766 on Build GRCh38
rs936127000 Chr.16: 3589767-3589767 on Build GRCh38
rs752013274 Chr.16: 3589657-3589657 on Build GRCh38
rs779981907 Chr.16: 3589763-3589763 on Build GRCh38
rs770785594 Chr.16: 3589478-3589478 on Build GRCh38
rs144720266 Chr.16: 3589583-3589583 on Build GRCh38
rs765606538 Chr.16: 3589522-3589522 on Build GRCh38
rs770811758 Chr.16: 3589540-3589540 on Build GRCh38
rs146079010 Chr.16: 3589441-3589441 on Build GRCh38
rs926938694 Chr.16: 3589492-3589492 on Build GRCh38
rs140892471 Chr.16: 3589397-3589397 on Build GRCh38
rs768867319 Chr.16: 3589701-3589701 on Build GRCh38
rs967380920 Chr.16: 3589576-3589576 on Build GRCh38
rs775548839 Chr.16: 3589717-3589719 on Build GRCh38
rs116781836 Chr.16: 3589675-3589675 on Build GRCh38
rs761812263 Chr.16: 3589349-3589349 on Build GRCh38
rs200310925 Chr.16: 3589605-3589605 on Build GRCh38
rs147600630 Chr.16: 3589687-3589687 on Build GRCh38
rs763731687 Chr.16: 3589732-3589732 on Build GRCh38
rs758320008 Chr.16: 3589761-3589761 on Build GRCh38
rs112022398 Chr.16: 3589597-3589597 on Build GRCh38
rs965203375 Chr.16: 3589426-3589426 on Build GRCh38
rs748999040 Chr.16: 3589473-3589473 on Build GRCh38
rs993089915 Chr.16: 3589453-3589453 on Build GRCh38
rs769920493 Chr.16: 3589350-3589350 on Build GRCh38
rs369714443 Chr.16: 3589699-3589699 on Build GRCh38
rs369846826 Chr.16: 3589524-3589524 on Build GRCh38
rs753464234 Chr.16: 3589735-3589735 on Build GRCh38
rs754092988 Chr.16: 3589412-3589412 on Build GRCh38
rs763708607 Chr.16: 3589667-3589667 on Build GRCh38
rs757513165 Chr.16: 3589507-3589507 on Build GRCh38
rs112596894 Chr.16: 3589770-3589770 on Build GRCh38
rs1028935047 Chr.16: 3589355-3589355 on Build GRCh38
rs762824802 Chr.16: 3589363-3589363 on Build GRCh38
rs771503932 Chr.16: 3589780-3589780 on Build GRCh38
rs115491049 Chr.16: 3589570-3589570 on Build GRCh38
rs756605546 Chr.16: 3589379-3589379 on Build GRCh38
rs764254888 Chr.16: 3589571-3589571 on Build GRCh38
rs751302297 Chr.16: 3589765-3589765 on Build GRCh38
rs138125792 Chr.16: 3589408-3589408 on Build GRCh38
rs376437029 Chr.16: 3589452-3589452 on Build GRCh38
rs891355584 Chr.16: 3589516-3589516 on Build GRCh38
rs766830918 Chr.16: 3589640-3589640 on Build GRCh38
rs781663547 Chr.16: 3589378-3589379 on Build GRCh38
rs200008765 Chr.16: 3589704-3589704 on Build GRCh38
rs752423507 Chr.16: 3589591-3589591 on Build GRCh38
rs773937495 Chr.16: 3589636-3589636 on Build GRCh38
rs149011965 Chr.16: 3589781-3589781 on Build GRCh38
rs749975141 Chr.16: 3589758-3589758 on Build GRCh38
rs368434737 Chr.16: 3589542-3589542 on Build GRCh38
rs759828401 Chr.16: 3589731-3589731 on Build GRCh38
rs748930384 Chr.16: 3589548-3589549 on Build GRCh38
rs374311917 Chr.16: 3589705-3589705 on Build GRCh38
rs773210318 Chr.16: 3589472-3589472 on Build GRCh38
rs201826765 Chr.16: 3589636-3589636 on Build GRCh38
rs371394594 Chr.16: 3589661-3589661 on Build GRCh38
rs142040192 Chr.16: 3589698-3589698 on Build GRCh38
rs747599109 Chr.16: 3589529-3589529 on Build GRCh38
rs140103960 Chr.16: 3589790-3589790 on Build GRCh38
rs376112909 Chr.16: 3589440-3589440 on Build GRCh38
rs778781581 Chr.16: 3589676-3589676 on Build GRCh38
rs774060802 Chr.16: 3589481-3589481 on Build GRCh38
rs748812906 Chr.16: 3589603-3589603 on Build GRCh38
rs1056055996 Chr.16: 3589370-3589370 on Build GRCh38
rs760555181 Chr.16: 3589724-3589725 on Build GRCh38
rs928993696 Chr.16: 3589679-3589679 on Build GRCh38
rs768829231 Chr.16: 3589618-3589618 on Build GRCh38
rs757202686 Chr.16: 3589604-3589604 on Build GRCh38
rs1040333386 Chr.16: 3589792-3589792 on Build GRCh38
rs532756081 Chr.16: 3589572-3589572 on Build GRCh38
rs778021365 Chr.16: 3589509-3589527 on Build GRCh38
rs566767778 Chr.16: 3589644-3589644 on Build GRCh38
rs766299864 Chr.16: 3589796-3589796 on Build GRCh38
rs374481298 Chr.16: 3589752-3589752 on Build GRCh38
rs756446820 Chr.16: 3589777-3589777 on Build GRCh38
rs761813549 Chr.16: 3589634-3589634 on Build GRCh38
rs759566186 Chr.16: 3589374-3589374 on Build GRCh38
rs767241703 Chr.16: 3589590-3589590 on Build GRCh38
rs762456970 Chr.16: 3589795-3589795 on Build GRCh38
rs768311081 Chr.16: 3589336-3589336 on Build GRCh38
rs771773313 Chr.16: 3589675-3589686 on Build GRCh38
rs757644807 Chr.16: 3589424-3589424 on Build GRCh38
rs771297945 Chr.16: 3589366-3589366 on Build GRCh38
rs770490824 Chr.16: 3589594-3589594 on Build GRCh38
rs377243383 Chr.16: 3589745-3589745 on Build GRCh38
rs755109579 Chr.16: 3589363-3589365 on Build GRCh38
rs771884438 Chr.16: 3589609-3589609 on Build GRCh38
rs775694333 Chr.16: 3589569-3589569 on Build GRCh38
rs373107728 Chr.16: 3589788-3589788 on Build GRCh38
rs772869326 Chr.16: 3589358-3589358 on Build GRCh38
rs574928535 Chr.16: 3589791-3589791 on Build GRCh38
rs367562934 Chr.16: 3589635-3589635 on Build GRCh38
rs148695476 Chr.16: 3589579-3589579 on Build GRCh38
rs760757846 Chr.16: 3589504-3589504 on Build GRCh38
rs745354595 Chr.16: 3589639-3589641 on Build GRCh38
rs766448056 Chr.16: 3589365-3589365 on Build GRCh38
rs763524714 Chr.16: 3589725-3589725 on Build GRCh38
rs983816841 Chr.16: 3589489-3589489 on Build GRCh38
rs953994627 Chr.16: 3589371-3589371 on Build GRCh38
rs141567438 Chr.16: 3589377-3589377 on Build GRCh38
rs772652088 Chr.16: 3589794-3589794 on Build GRCh38
rs541433695 Chr.16: 3589471-3589471 on Build GRCh38
rs767790750 Chr.16: 3589494-3589494 on Build GRCh38
rs200928592 Chr.16: 3589789-3589789 on Build GRCh38
rs745595506 Chr.16: 3589608-3589608 on Build GRCh38
rs772219842 Chr.16: 3589563-3589563 on Build GRCh38
rs188808130 Chr.16: 3589757-3589757 on Build GRCh38
rs533507487 Chr.16: 3589658-3589658 on Build GRCh38
rs140254478 Chr.16: 3589726-3589726 on Build GRCh38
rs777525901 Chr.16: 3589535-3589535 on Build GRCh38
rs371580654 Chr.16: 3589505-3589505 on Build GRCh38
rs777304392 Chr.16: 3589601-3589601 on Build GRCh38

View Full Product Details