Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24093964-24094464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCB
Entrez Gene ID
5579
Gene Name
protein kinase C beta
Gene Aliases
PKC-beta, PKCB, PRKCB1, PRKCB2
UniGene
Hs.460355
Gene Chromosome Location
Chr.16: 23835979-24220611 on Build GRCh38
Transcripts
NM_002738.6 NM_212535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4717700 c.716G>T p.R239I Chr.16: 24094192-24094192 on Build GRCh38
7301985 c.714A>G p.R238R Chr.16: 24094190-24094190 on Build GRCh38
1563175 c.784G>T p.G262W Chr.16: 24094260-24094260 on Build GRCh38
4257777 c.793G>A p.E265K Chr.16: 24094269-24094269 on Build GRCh38
4717702 c.716G>T p.R239I Chr.16: 24094192-24094192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778185299 Chr.16: 24094325-24094325 on Build GRCh38
rs749661878 Chr.16: 24094157-24094157 on Build GRCh38
rs565614049 Chr.16: 24094371-24094371 on Build GRCh38
rs986842078 Chr.16: 24094005-24094005 on Build GRCh38
rs1030260602 Chr.16: 24094427-24094427 on Build GRCh38
rs760228766 Chr.16: 24094349-24094349 on Build GRCh38
rs527666362 Chr.16: 24094395-24094395 on Build GRCh38
rs375014208 Chr.16: 24094139-24094139 on Build GRCh38
rs771532641 Chr.16: 24094347-24094347 on Build GRCh38
rs961967414 Chr.16: 24094077-24094077 on Build GRCh38
rs184224181 Chr.16: 24093994-24093994 on Build GRCh38
rs763829980 Chr.16: 24094226-24094226 on Build GRCh38
rs755036948 Chr.16: 24094292-24094292 on Build GRCh38
rs17847893 Chr.16: 24094377-24094377 on Build GRCh38
rs993552396 Chr.16: 24094424-24094424 on Build GRCh38
rs753635581 Chr.16: 24094232-24094232 on Build GRCh38
rs17847891 Chr.16: 24094190-24094190 on Build GRCh38
rs771584055 Chr.16: 24094181-24094181 on Build GRCh38
rs753204109 Chr.16: 24094304-24094304 on Build GRCh38
rs750947254 Chr.16: 24094336-24094336 on Build GRCh38
rs576580976 Chr.16: 24094350-24094350 on Build GRCh38
rs999067657 Chr.16: 24094355-24094355 on Build GRCh38
rs545711536 Chr.16: 24094365-24094365 on Build GRCh38
rs760639179 Chr.16: 24094214-24094214 on Build GRCh38
rs114093019 Chr.16: 24094178-24094178 on Build GRCh38
rs756684442 Chr.16: 24094281-24094281 on Build GRCh38
rs911786780 Chr.16: 24094025-24094025 on Build GRCh38
rs533575928 Chr.16: 24094036-24094036 on Build GRCh38
rs755165643 Chr.16: 24094132-24094132 on Build GRCh38
rs1030294650 Chr.16: 24094070-24094070 on Build GRCh38
rs7196765 Chr.16: 24094348-24094348 on Build GRCh38
rs777404333 Chr.16: 24094272-24094272 on Build GRCh38
rs1016139012 Chr.16: 24094367-24094367 on Build GRCh38
rs997050997 Chr.16: 24094431-24094431 on Build GRCh38
rs954926065 Chr.16: 24094103-24094103 on Build GRCh38
rs751743022 Chr.16: 24094288-24094288 on Build GRCh38
rs781448514 Chr.16: 24094295-24094295 on Build GRCh38
rs773646177 Chr.16: 24094155-24094155 on Build GRCh38
rs766610202 Chr.16: 24094274-24094274 on Build GRCh38
rs756663549 Chr.16: 24094322-24094322 on Build GRCh38
rs751095401 Chr.16: 24094154-24094154 on Build GRCh38
rs780602085 Chr.16: 24094404-24094404 on Build GRCh38
rs200005518 Chr.16: 24094164-24094164 on Build GRCh38
rs770589472 Chr.16: 24094177-24094177 on Build GRCh38
rs111892972 Chr.16: 24094435-24094435 on Build GRCh38
rs770240439 Chr.16: 24094151-24094151 on Build GRCh38
rs138174323 Chr.16: 24094343-24094343 on Build GRCh38
rs546408695 Chr.16: 24094326-24094326 on Build GRCh38
rs986359529 Chr.16: 24094135-24094135 on Build GRCh38
rs768076316 Chr.16: 24094327-24094327 on Build GRCh38
rs893295358 Chr.16: 24094376-24094376 on Build GRCh38
rs781741522 Chr.16: 24094136-24094136 on Build GRCh38
rs747105719 Chr.16: 24094122-24094122 on Build GRCh38

View Full Product Details