The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081.3 | NP_001072.2 | 43 - 44 | 6699 | 92 |
XM_011519708.2 | XP_011518010.1 | 43 - 44 | 7374 | 92 | |
XM_011519709.2 | XP_011518011.1 | 17 - 18 | 3062 | 92 | |
XM_011519710.2 | XP_011518012.1 | 17 - 18 | 2705 | 92 | |
XM_011519711.2 | XP_011518013.1 | 16 - 17 | 3386 | 92 | |
GenBank mRNA | AB209880.1 | - | 16 - 17 | 2766 | 92 |
AF034611.1 | - | 43 - 44 | 6673 | 92 |