Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCC1
Entrez Gene ID
1104
Gene Name
regulator of chromosome condensation 1
Gene Aliases
CHC1, RCC1-I, SNHG3-RCC1
Chromosome Location
Chr.1: 28505943 - 28539196 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048194.2 NP_001041659.1 5 - 6 623 68
NM_001048195.2 NP_001041660.1 5 - 6 581 68
NM_001048199.2 NP_001041664.1 6 - 7 705 68
NM_001269.4 NP_001260.1 4 - 5 530 68
NR_030725.1 7 - 8 848 68
NR_030726.1 7 - 8 844 68
GenBank mRNA AF498924.1 - 3 - 4 445 68
BC001589.1 - 3 - 4 467 68
BC010067.2 - 3 - 4 448 68
BC069198.1 - 5 - 6 588 68
BM546148.1 - 3 - 4 455 68
BQ436549.1 - 5 - 6 561 68
BU501614.1 - 6 - 7 658 68
CR407693.1 - 3 - 4 445 68
CV811053.1 - 5 - 6 552 68
KU177974.1 - 3 - 4 445 68
KU177975.1 - 4 - 5 538 68
KU177976.1 - 4 - 5 496 68
X12654.1 - 4 - 5 627 68

View Full Product Details