The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001879.5 | NP_001870.3 | 10 - 11 | 1693 | 90 |
XM_011512990.2 | XP_011511292.1 | 10 - 11 | 1687 | 90 | |
XM_017006870.1 | XP_016862359.1 | 9 - 10 | 1659 | 90 | |
GenBank mRNA | AL708781.1 | - | 2 - 3 | 177 | 90 |