Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EEF1A2
Entrez Gene ID
1917
Gene Name
eukaryotic translation elongation factor 1 alpha 2
Gene Aliases
EEF1AL, EF-1-alpha-2, EF1A, EIEE33, HS1, MRD38, STN, STNL
Chromosome Location
Chr.20: 63488012 - 63499315 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001958.3 NP_001949.1 4 - 5 950 87
GenBank mRNA AB209064.1 - 2 - 3 3190 87
AB451389.1 - 3 - 4 621 87
AK307536.1 - 4 - 5 718 87
BC000432.2 - 3 - 4 689 87
BC018855.2 - 2 - 3 331 87
BC110409.1 - 4 - 5 919 87
X70940.1 - 3 - 4 704 87

View Full Product Details