Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88809992-88810495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APRT
Entrez Gene ID
353
Gene Name
adenine phosphoribosyltransferase
Gene Aliases
AMP, APRTD
UniGene
Hs.28914
Gene Chromosome Location
Chr.16: 88809469-88811934 on Build GRCh38
Transcripts
NM_000485.2 NM_001030018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4486491 c.305C>T p.S102F Chr.16: 88810439-88810439 on Build GRCh38
1679333 c.333G>T p.E111D Chr.16: 88810137-88810137 on Build GRCh38
4063531 c.265C>T p.R89W Chr.16: 88810479-88810479 on Build GRCh38
974779 c.388C>T p.L130L Chr.16: 88810082-88810082 on Build GRCh38
7076145 c.260G>A p.R87Q Chr.16: 88810484-88810484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753423942 Chr.16: 88810399-88810399 on Build GRCh38
rs879169694 Chr.16: 88810268-88810273 on Build GRCh38
rs528515938 Chr.16: 88810116-88810116 on Build GRCh38
rs533009428 Chr.16: 88810142-88810142 on Build GRCh38
rs540232954 Chr.16: 88810385-88810385 on Build GRCh38
rs116389635 Chr.16: 88810188-88810188 on Build GRCh38
rs368062984 Chr.16: 88810423-88810423 on Build GRCh38
rs281860264 Chr.16: 88810451-88810451 on Build GRCh38
rs62048258 Chr.16: 88810025-88810025 on Build GRCh38
rs8191492 Chr.16: 88810212-88810212 on Build GRCh38
rs755069470 Chr.16: 88810289-88810289 on Build GRCh38
rs776240467 Chr.16: 88810458-88810464 on Build GRCh38
rs545344274 Chr.16: 88810243-88810243 on Build GRCh38
rs769047691 Chr.16: 88810044-88810044 on Build GRCh38
rs761430319 Chr.16: 88810473-88810474 on Build GRCh38
rs376141412 Chr.16: 88810163-88810163 on Build GRCh38
rs201944035 Chr.16: 88810124-88810124 on Build GRCh38
rs104894508 Chr.16: 88810141-88810141 on Build GRCh38
rs866574320 Chr.16: 88810406-88810406 on Build GRCh38
rs372253865 Chr.16: 88810468-88810468 on Build GRCh38
rs962326946 Chr.16: 88810280-88810280 on Build GRCh38
rs781444386 Chr.16: 88810312-88810312 on Build GRCh38
rs368691145 Chr.16: 88810377-88810377 on Build GRCh38
rs776948275 Chr.16: 88810111-88810111 on Build GRCh38
rs533045091 Chr.16: 88810170-88810170 on Build GRCh38
rs8191491 Chr.16: 88810229-88810229 on Build GRCh38
rs770055962 Chr.16: 88810426-88810426 on Build GRCh38
rs1041739461 Chr.16: 88810363-88810363 on Build GRCh38
rs982450640 Chr.16: 88810270-88810270 on Build GRCh38
rs760230688 Chr.16: 88810028-88810028 on Build GRCh38
rs960053989 Chr.16: 88810362-88810363 on Build GRCh38
rs201227087 Chr.16: 88810101-88810101 on Build GRCh38
rs1057424309 Chr.16: 88810378-88810378 on Build GRCh38
rs866738117 Chr.16: 88810274-88810274 on Build GRCh38
rs952402556 Chr.16: 88810260-88810260 on Build GRCh38
rs947408391 Chr.16: 88810207-88810207 on Build GRCh38
rs768699313 Chr.16: 88810438-88810438 on Build GRCh38
rs376129456 Chr.16: 88810085-88810085 on Build GRCh38
rs780413670 Chr.16: 88810465-88810465 on Build GRCh38
rs746631133 Chr.16: 88810032-88810032 on Build GRCh38
rs104894507 Chr.16: 88810450-88810450 on Build GRCh38
rs281860262 Chr.16: 88810447-88810447 on Build GRCh38
rs1021863023 Chr.16: 88810310-88810310 on Build GRCh38
rs774880744 Chr.16: 88810053-88810053 on Build GRCh38
rs763956969 Chr.16: 88810125-88810125 on Build GRCh38
rs745594160 Chr.16: 88810067-88810068 on Build GRCh38
rs771893788 Chr.16: 88810069-88810078 on Build GRCh38
rs200953386 Chr.16: 88810386-88810386 on Build GRCh38
rs139444461 Chr.16: 88810042-88810042 on Build GRCh38
rs932153375 Chr.16: 88810338-88810338 on Build GRCh38
rs764162455 Chr.16: 88810455-88810488 on Build GRCh38
rs747315064 Chr.16: 88810107-88810107 on Build GRCh38
rs753976270 Chr.16: 88810064-88810064 on Build GRCh38
rs755599871 Chr.16: 88810103-88810103 on Build GRCh38
rs748632043 Chr.16: 88810348-88810348 on Build GRCh38
rs866519045 Chr.16: 88810269-88810269 on Build GRCh38
rs1049078351 Chr.16: 88810209-88810209 on Build GRCh38
rs775353984 Chr.16: 88810123-88810123 on Build GRCh38
rs551418842 Chr.16: 88810443-88810443 on Build GRCh38
rs756911786 Chr.16: 88810098-88810098 on Build GRCh38
rs984020289 Chr.16: 88810390-88810390 on Build GRCh38
rs908129358 Chr.16: 88810157-88810157 on Build GRCh38
rs764924480 Chr.16: 88810474-88810474 on Build GRCh38
rs374197629 Chr.16: 88810059-88810059 on Build GRCh38
rs760635677 Chr.16: 88810128-88810128 on Build GRCh38
rs774658385 Chr.16: 88810063-88810063 on Build GRCh38
rs902033355 Chr.16: 88810089-88810089 on Build GRCh38
rs563575862 Chr.16: 88810456-88810456 on Build GRCh38
rs772379415 Chr.16: 88810105-88810105 on Build GRCh38
rs200019656 Chr.16: 88810154-88810154 on Build GRCh38
rs753415188 Chr.16: 88810470-88810470 on Build GRCh38
rs777336565 Chr.16: 88810031-88810031 on Build GRCh38
rs780755086 Chr.16: 88810100-88810100 on Build GRCh38
rs569463085 Chr.16: 88810251-88810251 on Build GRCh38
rs778476309 Chr.16: 88810407-88810407 on Build GRCh38
rs752084206 Chr.16: 88810408-88810408 on Build GRCh38
rs769819482 Chr.16: 88810178-88810178 on Build GRCh38
rs530663323 Chr.16: 88810458-88810458 on Build GRCh38
rs149329618 Chr.16: 88810184-88810184 on Build GRCh38
rs1002743528 Chr.16: 88810245-88810245 on Build GRCh38
rs376111313 Chr.16: 88810133-88810133 on Build GRCh38
rs281860263 Chr.16: 88810420-88810421 on Build GRCh38
rs368375344 Chr.16: 88810145-88810145 on Build GRCh38
rs8191489 Chr.16: 88810258-88810258 on Build GRCh38
rs572963308 Chr.16: 88810383-88810383 on Build GRCh38
rs200704114 Chr.16: 88810153-88810153 on Build GRCh38
rs8191490 Chr.16: 88810250-88810251 on Build GRCh38
rs947467388 Chr.16: 88810360-88810360 on Build GRCh38
rs201755177 Chr.16: 88810173-88810173 on Build GRCh38
rs543205392 Chr.16: 88810268-88810268 on Build GRCh38
rs151240811 Chr.16: 88810129-88810129 on Build GRCh38
rs202098006 Chr.16: 88810429-88810429 on Build GRCh38
rs533069674 Chr.16: 88810396-88810396 on Build GRCh38
rs765307269 Chr.16: 88810367-88810367 on Build GRCh38
rs376629164 Chr.16: 88810097-88810097 on Build GRCh38
rs8191488 Chr.16: 88810271-88810274 on Build GRCh38
rs746096749 Chr.16: 88810174-88810174 on Build GRCh38
rs754399799 Chr.16: 88810401-88810401 on Build GRCh38
rs71395327 Chr.16: 88810249-88810249 on Build GRCh38
rs1012725440 Chr.16: 88810254-88810254 on Build GRCh38
rs200417820 Chr.16: 88810474-88810474 on Build GRCh38
rs368883030 Chr.16: 88810055-88810055 on Build GRCh38
rs8191494 Chr.16: 88810108-88810108 on Build GRCh38
rs770492274 Chr.16: 88810041-88810041 on Build GRCh38
rs780098835 Chr.16: 88810428-88810428 on Build GRCh38
rs761643046 Chr.16: 88810191-88810191 on Build GRCh38
rs371083680 Chr.16: 88810180-88810180 on Build GRCh38
rs759652584 Chr.16: 88810073-88810073 on Build GRCh38
rs8191493 Chr.16: 88810185-88810185 on Build GRCh38
rs781677764 Chr.16: 88810422-88810422 on Build GRCh38
rs370665100 Chr.16: 88810118-88810118 on Build GRCh38
rs373048958 Chr.16: 88810056-88810056 on Build GRCh38
rs200229196 Chr.16: 88810264-88810265 on Build GRCh38
rs75205792 Chr.16: 88810094-88810094 on Build GRCh38
rs771403483 Chr.16: 88810161-88810161 on Build GRCh38
rs35095508 Chr.16: 88810106-88810106 on Build GRCh38
rs775204282 Chr.16: 88810073-88810082 on Build GRCh38
rs542789654 Chr.16: 88810104-88810104 on Build GRCh38
rs561088237 Chr.16: 88810357-88810357 on Build GRCh38
rs767177754 Chr.16: 88810136-88810136 on Build GRCh38
rs1011477101 Chr.16: 88810084-88810084 on Build GRCh38
rs750053169 Chr.16: 88810140-88810140 on Build GRCh38
rs986292805 Chr.16: 88810339-88810339 on Build GRCh38
rs745324089 Chr.16: 88810043-88810043 on Build GRCh38
rs763991694 Chr.16: 88810095-88810095 on Build GRCh38
rs757760484 Chr.16: 88810411-88810411 on Build GRCh38
rs760967219 Chr.16: 88810057-88810057 on Build GRCh38

View Full Product Details