The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172357.1 | NP_001165828.1 | 1 - 2 | 202 | 73 |
NM_001172410.1 | NP_001165881.1 | 1 - 2 | 202 | 73 | |
NM_001317778.1 | NP_001304707.1 | 1 - 2 | 202 | 73 | |
NM_001317780.1 | NP_001304709.1 | 1 - 2 | 202 | 73 | |
NM_003018.3 | NP_003009.2 | 1 - 2 | 202 | 73 | |
GenBank mRNA | AK058094.1 | - | 4 - 5 | 894 | 73 |
AK298119.1 | - | 1 - 2 | 73 | 73 | |
AK315742.1 | - | 1 - 2 | 76 | 73 | |
BC005913.1 | - | 1 - 2 | 75 | 73 | |
BG535994.1 | - | 1 - 2 | 79 | 73 | |
BG569816.1 | - | 1 - 2 | 79 | 73 | |
BI524154.1 | - | 1 - 2 | 59 | 73 | |
DQ884411.1 | - | 1 - 2 | 44 | 73 | |
J03517.1 | - | 1 - 2 | 71 | 73 | |
J03553.1 | - | 1 - 2 | 218 | 73 | |
KU178329.1 | - | 1 - 2 | 44 | 73 | |
KU178330.1 | - | 1 - 2 | 44 | 73 |