The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003369.3 | NP_003360.2 | 11 - 12 | 1305 | 68 |
XM_011545226.2 | XP_011543528.1 | 11 - 12 | 1620 | 68 | |
XM_011545227.2 | XP_011543529.1 | 11 - 12 | 1620 | 68 | |
XM_011545228.2 | XP_011543530.1 | 11 - 12 | 1620 | 68 | |
XM_017018224.1 | XP_016873713.1 | 9 - 10 | 1059 | 68 | |
XM_017018225.1 | XP_016873714.1 | 9 - 10 | 1059 | 68 | |
XM_017018226.1 | XP_016873715.1 | 11 - 12 | 1620 | 68 | |
GenBank mRNA | AB012958.1 | - | 11 - 12 | 1242 | 68 |
AK095352.1 | - | 5 - 6 | 882 | 68 | |
AK222772.1 | - | 11 - 12 | 1196 | 68 | |
AK291011.1 | - | 11 - 12 | 1283 | 68 | |
AK293782.1 | - | 11 - 12 | 1046 | 68 | |
AK296871.1 | - | 5 - 6 | 425 | 68 | |
BC064837.1 | - | 11 - 12 | 1215 | 68 |