The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000136.2 | NP_000127.2 | 10 - 11 | 1258 | 82 |
NM_001243743.1 | NP_001230672.1 | 10 - 11 | 1200 | 82 | |
NM_001243744.1 | NP_001230673.1 | 10 - 11 | 1258 | 82 | |
XM_005251802.3 | XP_005251859.1 | 4 - 5 | 1360 | 82 | |
XM_006717001.2 | XP_006717064.1 | 9 - 10 | 1075 | 82 | |
XM_006717002.3 | XP_006717065.1 | 10 - 11 | 1240 | 82 | |
XM_011518365.2 | XP_011516667.1 | 10 - 11 | 1233 | 82 | |
XM_011518366.2 | XP_011516668.1 | 10 - 11 | 1240 | 82 | |
XM_011518367.2 | XP_011516669.1 | 11 - 12 | 1370 | 82 | |
XM_017014452.1 | XP_016869941.1 | 11 - 12 | 1385 | 82 | |
XM_017014453.1 | XP_016869942.1 | 11 - 12 | 1332 | 82 | |
XM_017014454.1 | XP_016869943.1 | 10 - 11 | 1259 | 82 | |
GenBank mRNA | AK222871.1 | - | 10 - 11 | 1220 | 82 |
AK312548.1 | - | 10 - 11 | 1123 | 82 | |
BC015748.1 | - | 10 - 11 | 1248 | 82 | |
KU178051.1 | - | 9 - 10 | 996 | 82 | |
KU178052.1 | - | 10 - 11 | 1141 | 82 | |
X66893.1 | - | 10 - 11 | 1251 | 82 | |
X66894.1 | - | 10 - 11 | 1169 | 82 |