Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEFV
Entrez Gene ID
4210
Gene Name
Mediterranean fever
Gene Aliases
FMF, MEF, TRIM20
Chromosome Location
Chr.16: 3242028 - 3256776 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000243.2 NP_000234.1 2 - 3 954 64
XM_017023236.1 XP_016878725.1 2 - 3 1095 64
GenBank mRNA AF018080.1 - 2 - 3 955 64
AK298660.1 - 2 - 3 948 64
BC101511.1 - 2 - 3 939 64
BC101537.1 - 2 - 3 939 64
Y14441.1 - 2 - 3 943 64

View Full Product Details