Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCCA
Entrez Gene ID
5095
Gene Name
propionyl-CoA carboxylase alpha subunit
Gene Aliases
-
Chromosome Location
Chr.13: 100089015 - 100530437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000282.3 NP_000273.2 20 - 21 1953 64
NM_001127692.2 NP_001121164.1 19 - 20 1875 64
NM_001178004.1 NP_001171475.1 20 - 21 1953 64
XM_005254059.2 XP_005254116.1 20 - 21 1948 64
XM_011521093.1 XP_011519395.1 20 - 21 1948 64
XM_017020609.1 XP_016876098.1 20 - 21 1861 64
XM_017020614.1 XP_016876103.1 19 - 20 1799 64
XM_017020617.1 XP_016876106.1 16 - 17 1532 64
GenBank mRNA AF385926.1 - 20 - 21 1885 64
AK225288.1 - 20 - 21 1853 64
AK225299.1 - 20 - 21 1851 64
AK298318.1 - 20 - 21 1875 64
AK315503.1 - 20 - 21 1851 64
BC000140.2 - 20 - 21 1825 64
S79219.1 - 4 - 5 345 64
X14608.1 - 20 - 21 1820 64

View Full Product Details