Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX6
Entrez Gene ID
5190
Gene Name
peroxisomal biogenesis factor 6
Gene Aliases
HMLR2, PAF-2, PAF2, PBD4A, PDB4B, PXAAA1
Chromosome Location
Chr.6: 42963873 - 42979243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000287.3 NP_000278.3 3 - 4 1226 86
NM_001316313.1 NP_001303242.1 3 - 4 962 86
NR_133009.1 3 - 4 1226 86
GenBank mRNA AB051076.1 - 3 - 4 1133 86
AB051077.1 - 3 - 4 869 86
AB051078.1 - 3 - 4 1133 86
AB208906.1 - 3 - 4 1186 86
AK225704.1 - 3 - 4 1164 86
AK314237.1 - 3 - 4 1162 86
BC033487.1 - 3 - 4 1141 86
BC048331.1 - 3 - 4 1149 86
D83703.1 - 3 - 4 1203 86

View Full Product Details