The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000597.2 | NP_000588.2 | 1 - 2 | 574 | 70 |
GenBank mRNA | A09809.1 | - | 1 - 2 | 573 | 70 |
AB209509.1 | - | 1 - 2 | 549 | 70 | |
AL559513.3 | - | 1 - 2 | 582 | 70 | |
BC004312.1 | - | 1 - 2 | 548 | 70 | |
BC009902.1 | - | 1 - 2 | 550 | 70 | |
BC012769.1 | - | 1 - 2 | 548 | 70 | |
BC071967.1 | - | 1 - 2 | 561 | 70 | |
M35410.1 | - | 1 - 2 | 517 | 70 | |
X16302.1 | - | 1 - 2 | 571 | 70 |