The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000670.4 | NP_000661.2 | 6 - 7 | 921 | 60 |
NM_001306171.1 | NP_001293100.1 | 7 - 8 | 1069 | 60 | |
NM_001306172.1 | NP_001293101.1 | 7 - 8 | 1061 | 60 | |
XM_017007713.1 | XP_016863202.1 | 7 - 8 | 1092 | 60 | |
XM_017007714.1 | XP_016863203.1 | 7 - 8 | 1100 | 60 | |
XM_017007715.1 | XP_016863204.1 | 6 - 7 | 953 | 60 | |
GenBank mRNA | AK290835.1 | - | 6 - 7 | 920 | 60 |
AK295556.1 | - | 7 - 8 | 1068 | 60 | |
AK313649.1 | - | 6 - 7 | 920 | 60 | |
BC022319.1 | - | 6 - 7 | 920 | 60 | |
CD014080.1 | - | 7 - 8 | 1039 | 60 | |
EU668317.1 | - | 6 - 7 | 906 | 60 | |
M15943.1 | - | 6 - 7 | 906 | 60 |