Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTR2B
Entrez Gene ID
3357
Gene Name
5-hydroxytryptamine receptor 2B
Gene Aliases
5-HT(2B), 5-HT-2B, 5-HT2B
Chromosome Location
Chr.2: 231108230 - 231125778 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000867.4 NP_000858.3 2 - 3 857 70
NM_001320758.1 NP_001307687.1 2 - 3 474 70
XM_006712482.2 XP_006712545.1 2 - 3 1054 70
GenBank mRNA AK313741.1 - 2 - 3 789 70
AY136751.1 - 1 - 2 352 70
BC063123.1 - 2 - 3 861 70
X77307.1 - 1 - 2 407 70
Z36748.1 - 1 - 2 398 70

View Full Product Details