The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022552.4 | NP_072046.2 | 4 - 5 | 719 | 96 |
NM_175629.2 | NP_783328.1 | 4 - 5 | 790 | 96 | |
NR_135490.1 | 4 - 5 | 790 | 96 | ||
XM_005264175.4 | XP_005264232.1 | 4 - 5 | 653 | 96 | |
XM_011532662.2 | XP_011530964.1 | 2 - 3 | 655 | 96 | |
XM_011532664.2 | XP_011530966.1 | 4 - 5 | 730 | 96 | |
XM_017003526.1 | XP_016859015.1 | 4 - 5 | 761 | 96 | |
GenBank mRNA | AB208833.1 | - | 4 - 5 | 689 | 96 |
AF067972.2 | - | 4 - 5 | 681 | 96 | |
AF331856.1 | - | 4 - 5 | 669 | 96 | |
BC043617.1 | - | 4 - 5 | 689 | 96 |