The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014322.2 | NP_055137.2 | 1 - 2 | 521 | 60 |
GenBank mRNA | AF140242.1 | - | 1 - 2 | 431 | 60 |
AF303588.1 | - | 1 - 2 | 478 | 60 | |
BC036773.1 | - | 1 - 2 | 473 | 60 | |
BC062622.1 | - | 1 - 2 | 462 | 60 |