The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004123.2 | NP_004114.1 | 2 - 3 | 189 | 78 |
GenBank mRNA | BC069100.1 | - | 2 - 3 | 189 | 78 |
BC069663.1 | - | 2 - 3 | 189 | 78 | |
BC069686.1 | - | 2 - 3 | 189 | 78 | |
BC069746.1 | - | 2 - 3 | 189 | 78 | |
BC096146.1 | - | 2 - 3 | 188 | 78 | |
BC096147.1 | - | 2 - 3 | 188 | 78 | |
BC096148.3 | - | 2 - 3 | 188 | 78 | |
BC096149.3 | - | 2 - 3 | 188 | 78 | |
E02006.1 | - | 2 - 3 | 191 | 78 | |
M18185.1 | - | 2 - 3 | 189 | 78 |