Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSNK1G2
Entrez Gene ID
1455
Gene Name
casein kinase 1 gamma 2
Gene Aliases
CK1g2
Chromosome Location
Chr.19: 1941149 - 1981338 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319.6 NP_001310.3 2 - 3 707 74
XM_005259498.1 XP_005259555.1 2 - 3 719 74
XM_005259499.3 XP_005259556.1 2 - 3 526 74
XM_005259500.1 XP_005259557.1 2 - 3 719 74
XM_017026297.1 XP_016881786.1 2 - 3 593 74
GenBank mRNA AB451278.1 - 1 - 2 184 74
AB451410.1 - 1 - 2 184 74
BC018693.2 - 2 - 3 680 74
BC018699.2 - 2 - 3 680 74
BC020972.1 - 1 - 2 237 74
BT009922.1 - 1 - 2 184 74
U89896.1 - 1 - 2 423 74

View Full Product Details