The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002035.2 | NP_002026.1 | 9 - 10 | 1039 | 79 |
XM_017025690.1 | XP_016881179.1 | 9 - 10 | 1244 | 79 | |
GenBank mRNA | AK025120.1 | - | 9 - 10 | 972 | 79 |
AK126679.1 | - | 3 - 4 | 2312 | 79 | |
AK297670.1 | - | 8 - 9 | 778 | 79 | |
AK312360.1 | - | 9 - 10 | 967 | 79 | |
BC008797.2 | - | 9 - 10 | 1018 | 79 | |
BT006782.1 | - | 9 - 10 | 884 | 79 | |
X63657.1 | - | 9 - 10 | 991 | 79 |