The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323370.1 | NP_001310299.1 | 4 - 5 | 878 | 81 |
NM_003881.3 | NP_003872.1 | 3 - 4 | 1082 | 81 | |
XM_017028116.1 | XP_016883605.1 | 2 - 3 | 393 | 81 | |
GenBank mRNA | AF074604.1 | - | 3 - 4 | 541 | 81 |
AF083500.1 | - | 3 - 4 | 543 | 81 | |
AF100780.1 | - | 3 - 4 | 690 | 81 | |
AK129660.1 | - | 1 - 2 | 1339 | 81 | |
AK313853.1 | - | 3 - 4 | 558 | 81 | |
AY358915.1 | - | 3 - 4 | 544 | 81 | |
BC017782.2 | - | 3 - 4 | 681 | 81 | |
BC064379.1 | - | 3 - 4 | 545 | 81 |