Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBLB
Entrez Gene ID
868
Gene Name
Cbl proto-oncogene B
Gene Aliases
Cbl-b, Nbla00127, RNF56
Chromosome Location
Chr.3: 105655461 - 105869552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321786.1 NP_001308715.1 6 - 7 1072 98
NM_001321788.1 NP_001308717.1 6 - 7 991 98
NM_001321789.1 NP_001308718.1 6 - 7 1010 98
NM_001321790.1 NP_001308719.1 6 - 7 945 98
NM_001321791.1 NP_001308720.1 6 - 7 991 98
NM_001321793.1 NP_001308722.1 6 - 7 1167 98
NM_001321794.1 NP_001308723.1 6 - 7 991 98
NM_001321795.1 NP_001308724.1 6 - 7 1167 98
NM_001321796.1 NP_001308725.1 5 - 6 844 98
NM_001321797.1 NP_001308726.1 6 - 7 991 98
NM_001321798.1 NP_001308727.1 6 - 7 1167 98
NM_001321799.1 NP_001308728.1 5 - 6 1020 98
NM_001321806.1 NP_001308735.1 5 - 6 834 98
NM_001321807.1 NP_001308736.1 7 - 8 1311 98
NM_001321808.1 NP_001308737.1 7 - 8 1135 98
NM_001321811.1 NP_001308740.1 5 - 6 1010 98
NM_001321813.1 NP_001308742.1 7 - 8 1048 98
NM_001321816.1 NP_001308745.1 5 - 6 1010 98
NM_001321820.1 NP_001308749.1 7 - 8 1135 98
NM_001321822.1 NP_001308751.1 6 - 7 1167 98
NM_170662.4 NP_733762.2 6 - 7 1167 98
NR_135806.1 7 - 8 1299 98
NR_135807.1 7 - 8 1215 98
NR_135808.1 6 - 7 1167 98
NR_135809.1 6 - 7 1167 98
NR_135810.1 6 - 7 945 98
NR_135811.1 6 - 7 991 98
NR_135812.1 6 - 7 887 98
XM_011513257.1 XP_011511559.1 6 - 7 987 98
XM_011513259.2 XP_011511561.2 6 - 7 1112 98
XM_017007395.1 XP_016862884.1 6 - 7 939 98
XM_017007396.1 XP_016862885.1 6 - 7 1110 98
XM_017007397.1 XP_016862886.1 5 - 6 964 98
XM_017007398.1 XP_016862887.1 5 - 6 790 98
XM_017007399.1 XP_016862888.1 5 - 6 1015 98
XM_017007400.1 XP_016862889.1 5 - 6 964 98
GenBank mRNA AK292792.1 - 6 - 7 1108 98
AK302533.1 - 6 - 7 945 98
AK302892.1 - 6 - 7 920 98
AK316276.1 - 6 - 7 925 98
BC032851.2 - 6 - 7 1108 98
DQ349203.1 - 5 - 6 845 98
DQ349204.1 - 5 - 6 845 98
U26710.1 - 6 - 7 1167 98
U26711.1 - 6 - 7 1167 98
U26712.1 - 6 - 7 1167 98

View Full Product Details