Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTN1
Entrez Gene ID
9423
Gene Name
netrin 1
Gene Aliases
NTN1L
Chromosome Location
Chr.17: 9021542 - 9244000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004822.2 NP_004813.2 4 - 5 1466 121
XM_006721595.3 XP_006721658.1 4 - 5 1566 121
GenBank mRNA GU727649.1 - 3 - 4 1366 121
U75586.1 - 4 - 5 1468 121

View Full Product Details