The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000836.2 | NP_000827.2 | 10 - 11 | 2340 | 74 |
XM_011526872.1 | XP_011525174.1 | 9 - 10 | 2508 | 74 | |
GenBank mRNA | U77783.1 | - | 10 - 11 | 2340 | 74 |