The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018009.3 | NP_001018009.2 | 3 - 4 | 435 | 90 |
NM_004844.4 | NP_004835.2 | 3 - 4 | 554 | 90 | |
XM_011534251.2 | XP_011532553.1 | 3 - 4 | 366 | 90 | |
XM_011534253.1 | XP_011532555.1 | 3 - 4 | 515 | 90 | |
XM_017007521.1 | XP_016863010.1 | 3 - 4 | 349 | 90 | |
XM_017007523.1 | XP_016863012.1 | 3 - 4 | 770 | 90 | |
XM_017007524.1 | XP_016863013.1 | 3 - 4 | 302 | 90 | |
XM_017007525.1 | XP_016863014.1 | 3 - 4 | 216 | 90 | |
GenBank mRNA | AB005047.1 | - | 3 - 4 | 306 | 90 |
AK313167.1 | - | 3 - 4 | 512 | 90 | |
AL117422.1 | - | 3 - 4 | 511 | 90 | |
AW673562.1 | - | 1 - 2 | 119 | 90 | |
BC006169.1 | - | 3 - 4 | 409 | 90 | |
BC010123.2 | - | 3 - 4 | 485 | 90 | |
BC023195.1 | - | 3 - 4 | 226 | 90 | |
BG475051.1 | - | - | 368 | 90 | |
BP308415.1 | - | 3 - 4 | 553 | 90 |