The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005842.3 | NP_005833.1 | 1 - 2 | 497 | 86 |
GenBank mRNA | AF039843.1 | - | 1 - 2 | 342 | 86 |
BC015745.1 | - | 1 - 2 | 58 | 86 | |
CX866409.1 | - | 1 - 2 | 497 | 86 |