The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303245.1 | NP_001290174.1 | 1 - 2 | 173 | 74 |
NM_001303246.1 | NP_001290175.1 | 1 - 2 | 173 | 74 | |
NM_006506.3 | NP_006497.2 | 1 - 2 | 173 | 74 | |
GenBank mRNA | AF115573.1 | - | 1 - 2 | 136 | 74 |
AK292016.1 | - | 1 - 2 | 171 | 74 | |
AK301015.1 | - | 1 - 2 | 171 | 74 | |
D78155.1 | - | 1 - 2 | 136 | 74 | |
D82880.1 | - | 1 - 2 | 136 | 74 |