Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- KLRK1
- Entrez Gene ID
-
22914
- Gene Name
- killer cell lectin like receptor K1
- Gene Aliases
-
CD314, D12S2489E, KLR, NKG2-D, NKG2D
- Chromosome Location
-
Chr.12: 10372353 - 10390054 on Build GRCh38
- Gene Symbol
- KLRC4-KLRK1
- Entrez Gene ID
-
100528032
- Gene Name
- KLRC4-KLRK1 readthrough
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 10372353 - 10410146 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF439512.1 |
- |
2 - 3 |
128 |
85 |
X54870.1 |
- |
3 - 4 |
276 |
85 |
View Full Product Details