The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012242.2 | NP_036374.1 | 2 - 3 | 559 | 68 |
GenBank mRNA | AF127563.1 | - | 2 - 3 | 544 | 68 |
AF177394.1 | - | 2 - 3 | 507 | 68 | |
AK314902.1 | - | 2 - 3 | 557 | 68 | |
AY359005.1 | - | 2 - 3 | 542 | 68 | |
BC001539.2 | - | 2 - 3 | 544 | 68 | |
JN966744.1 | - | 2 - 3 | 588 | 68 |