The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001975.1 | NP_001001975.1 | 1 - 2 | 241 | 73 |
NM_001687.4 | NP_001678.1 | 1 - 2 | 241 | 73 | |
GenBank mRNA | BC002389.2 | - | 1 - 2 | 241 | 73 |
BC004426.1 | - | 1 - 2 | 241 | 73 | |
BC018079.1 | - | 1 - 2 | 143 | 73 | |
BC018817.2 | - | 1 - 2 | 241 | 73 | |
CR542218.1 | - | 1 - 2 | 140 | 73 | |
S87916.1 | - | 1 - 2 | 223 | 73 | |
S87918.1 | - | 1 - 2 | 223 | 73 | |
X63422.1 | - | 1 - 2 | 223 | 73 | |
X63423.1 | - | 1 - 2 | 223 | 73 |