The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166292.1 | NP_001159764.1 | 19 - 20 | 3126 | 76 |
NM_003738.4 | NP_003729.3 | 19 - 20 | 3126 | 76 | |
GenBank mRNA | AF087651.1 | - | 19 - 20 | 3114 | 76 |
AF091501.1 | - | 19 - 20 | 3114 | 76 | |
AF119569.1 | - | 19 - 20 | 3126 | 76 | |
AY358555.1 | - | 19 - 20 | 3114 | 76 | |
AY359016.1 | - | 19 - 20 | 3118 | 76 | |
BC143712.1 | - | 18 - 19 | 2983 | 76 |