The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003742.2 | NP_003733.2 | 21 - 22 | 2736 | 63 |
XM_006712817.3 | XP_006712880.1 | 21 - 22 | 2848 | 63 | |
XM_011512077.2 | XP_011510379.1 | 21 - 22 | 2908 | 63 | |
XM_011512078.2 | XP_011510380.1 | 21 - 22 | 2908 | 63 | |
XM_011512081.2 | XP_011510383.1 | 8 - 9 | 1299 | 63 | |
XM_017005165.1 | XP_016860654.1 | 21 - 22 | 2910 | 63 | |
XM_017005166.1 | XP_016860655.1 | 15 - 16 | 2034 | 63 | |
XM_017005167.1 | XP_016860656.1 | 11 - 12 | 1592 | 63 | |
GenBank mRNA | AF091582.1 | - | 21 - 22 | 2736 | 63 |
AF136523.1 | - | 21 - 22 | 2737 | 63 | |
AK302540.1 | - | 8 - 9 | 1187 | 63 | |
AK303050.1 | - | 8 - 9 | 1299 | 63 |