The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001058.3 | NP_001049.1 | 2 - 3 | 1173 | 67 |
NM_015727.2 | NP_056542.1 | 2 - 3 | 1173 | 67 | |
GenBank mRNA | AK289765.1 | - | 2 - 3 | 1142 | 67 |
AY462098.1 | - | 2 - 3 | 588 | 67 | |
BC074911.2 | - | 2 - 3 | 610 | 67 | |
BC074912.2 | - | 2 - 3 | 610 | 67 | |
BC095528.1 | - | 2 - 3 | 585 | 67 | |
M74290.1 | - | 2 - 3 | 798 | 67 | |
M76675.1 | - | 2 - 3 | 588 | 67 | |
M81797.1 | - | 2 - 3 | 632 | 67 | |
M84425.1 | - | 2 - 3 | 710 | 67 | |
M84426.1 | - | 2 - 3 | 710 | 67 | |
S62045.1 | - | 2 - 3 | 652 | 67 |