The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290104.1 | NP_001277033.1 | 5 - 6 | 813 | 82 |
NM_003460.2 | NP_003451.1 | 5 - 6 | 813 | 82 | |
XM_011545942.2 | XP_011544244.1 | 4 - 5 | 342 | 82 | |
GenBank mRNA | AK293805.1 | - | 4 - 5 | 357 | 82 |
AK308820.1 | - | 4 - 5 | 357 | 82 | |
AK310454.1 | - | 5 - 6 | 813 | 82 | |
AK313003.1 | - | 4 - 5 | 355 | 82 | |
BC096304.3 | - | 4 - 5 | 330 | 82 | |
BC096305.3 | - | 4 - 5 | 330 | 82 | |
BC096306.1 | - | 4 - 5 | 330 | 82 | |
BC096307.3 | - | 4 - 5 | 330 | 82 | |
M90366.1 | - | 4 - 5 | 330 | 82 |