The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042544.1 | NP_001036009.1 | 24 - 25 | 3275 | 71 |
NM_001042545.1 | NP_001036010.1 | 21 - 22 | 3091 | 71 | |
NM_003573.2 | NP_003564.2 | 24 - 25 | 3164 | 71 | |
XM_011527376.2 | XP_011525678.1 | 23 - 24 | 3418 | 71 | |
XM_011527377.2 | XP_011525679.1 | 24 - 25 | 3323 | 71 | |
XM_011527378.2 | XP_011525680.1 | 24 - 25 | 3323 | 71 | |
XM_011527379.1 | XP_011525681.1 | 20 - 21 | 3226 | 71 | |
XM_011527380.2 | XP_011525682.1 | 23 - 24 | 3197 | 71 | |
XM_011527382.2 | XP_011525684.1 | 23 - 24 | 3080 | 71 | |
XM_011527383.2 | XP_011525685.1 | 24 - 25 | 3323 | 71 | |
XM_011527385.2 | XP_011525687.1 | 23 - 24 | 3191 | 71 | |
XM_011527387.1 | XP_011525689.1 | 24 - 25 | 3423 | 71 | |
XM_017027352.1 | XP_016882841.1 | 23 - 24 | 3191 | 71 | |
GenBank mRNA | AF051344.2 | - | 21 - 22 | 3091 | 71 |
AF051345.2 | - | 24 - 25 | 3275 | 71 | |
AF054501.1 | - | 3 - 4 | 361 | 71 | |
AK128191.1 | - | 10 - 11 | 1273 | 71 | |
Y13622.1 | - | 23 - 24 | 3164 | 71 |