The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166695.2 | NP_001160167.1 | 4 - 5 | 998 | 60 |
NM_001289939.1 | NP_001276868.1 | 3 - 4 | 860 | 60 | |
NM_004172.4 | NP_004163.3 | 4 - 5 | 998 | 60 | |
XM_005248342.2 | XP_005248399.1 | 5 - 6 | 851 | 60 | |
XM_011514084.2 | XP_011512386.1 | 5 - 6 | 849 | 60 | |
GenBank mRNA | AK293886.1 | - | 3 - 4 | 586 | 60 |
AK295032.1 | - | 3 - 4 | 394 | 60 | |
AK312304.1 | - | 4 - 5 | 766 | 60 | |
AY954110.1 | - | 3 - 4 | 522 | 60 | |
BC037310.2 | - | 4 - 5 | 766 | 60 | |
D26443.1 | - | 4 - 5 | 700 | 60 | |
L19158.1 | - | 4 - 5 | 661 | 60 | |
U03504.1 | - | 3 - 4 | 552 | 60 |