The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004345.4 | NP_004336.3 | 1 - 2 | 365 | 86 |
GenBank mRNA | AY162210.1 | - | 1 - 2 | 200 | 86 |
AY251531.1 | - | 1 - 2 | 203 | 86 | |
BC055089.1 | - | 1 - 2 | 242 | 86 | |
BU074012.1 | - | 1 - 2 | 368 | 86 | |
CR457083.1 | - | 1 - 2 | 200 | 86 | |
U19970.1 | - | 1 - 2 | 215 | 86 | |
X89658.1 | - | 1 - 2 | 235 | 86 | |
Z38026.1 | - | 1 - 2 | 211 | 86 |