The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282597.2 | NP_001269526.1 | 16 - 17 | 2571 | 95 |
NM_001282598.1 | NP_001269527.1 | 16 - 17 | 2424 | 95 | |
NM_001282599.1 | NP_001269528.1 | 10 - 11 | 1512 | 95 | |
NM_001282600.1 | NP_001269529.1 | 10 - 11 | 1455 | 95 | |
NM_001320810.1 | NP_001307739.1 | 10 - 11 | 1455 | 95 | |
NM_004389.3 | NP_004380.2 | 16 - 17 | 2571 | 95 | |
XM_011532555.2 | XP_011530857.1 | 16 - 17 | 2418 | 95 | |
XM_011532556.2 | XP_011530858.1 | 17 - 18 | 2614 | 95 | |
XM_011532557.2 | XP_011530859.1 | 8 - 9 | 1160 | 95 | |
XM_017003403.1 | XP_016858892.1 | 17 - 18 | 2614 | 95 | |
XM_017003404.1 | XP_016858893.1 | 16 - 17 | 2509 | 95 | |
XM_017003405.1 | XP_016858894.1 | 16 - 17 | 2509 | 95 | |
GenBank mRNA | AK127226.1 | - | 20 - 21 | 3017 | 95 |
AK294362.1 | - | 2 - 3 | 536 | 95 | |
AK295493.1 | - | 10 - 11 | 1442 | 95 | |
AK303035.1 | - | 16 - 17 | 2424 | 95 | |
BC040458.1 | - | 15 - 16 | 2243 | 95 | |
BC052996.1 | - | 16 - 17 | 2505 | 95 | |
BX537769.1 | - | 10 - 11 | 1512 | 95 | |
BX648368.1 | - | 3 - 4 | 384 | 95 | |
HQ589336.1 | - | 10 - 11 | 1443 | 95 | |
M94151.2 | - | 15 - 16 | 2322 | 95 |