The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004553.4 | NP_004544.1 | 3 - 4 | 347 | 102 |
GenBank mRNA | AF044959.1 | - | 3 - 4 | 321 | 102 |
AK022503.1 | - | 3 - 4 | 330 | 102 | |
BC038664.1 | - | 3 - 4 | 326 | 102 | |
BC046155.2 | - | 3 - 4 | 316 | 102 | |
BG706812.1 | - | 3 - 4 | 336 | 102 | |
BM172493.1 | - | 3 - 4 | 202 | 102 | |
BM759028.1 | - | 3 - 4 | 334 | 102 | |
BM764340.1 | - | 3 - 4 | 334 | 102 | |
CN285188.1 | - | 3 - 4 | 301 | 102 | |
CR456880.1 | - | 3 - 4 | 311 | 102 |