The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004751.2 | NP_004742.1 | 2 - 3 | 388 | 80 |
GenBank mRNA | AF102542.1 | - | 1 - 2 | 292 | 80 |
AK312852.1 | - | 1 - 2 | 171 | 80 | |
BC017032.1 | - | 2 - 3 | 247 | 80 | |
EF152283.1 | - | 2 - 3 | 529 | 80 |