The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004798.3 | NP_004789.1 | 2 - 3 | 1587 | 68 |
GenBank mRNA | AB002357.1 | - | 2 - 3 | 1574 | 68 |
AK098431.1 | - | 1 - 2 | 142 | 68 | |
AK299877.1 | - | 2 - 3 | 510 | 68 | |
BC136310.1 | - | 1 - 2 | 1480 | 68 | |
BC136311.1 | - | 1 - 2 | 1480 | 68 |