The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271440.1 | NP_001258369.1 | 3 - 4 | 353 | 94 |
NM_001271441.1 | NP_001258370.1 | 3 - 4 | 353 | 94 | |
NM_004928.2 | NP_004919.1 | 3 - 4 | 353 | 94 | |
XM_017028470.1 | XP_016883959.1 | 3 - 4 | 364 | 94 | |
GenBank mRNA | BC031300.1 | - | 3 - 4 | 344 | 94 |
BC072012.1 | - | 3 - 4 | 326 | 94 | |
BM761336.1 | - | 3 - 4 | 353 | 94 | |
U84569.1 | - | 3 - 4 | 325 | 94 | |
Y11392.1 | - | 3 - 4 | 213 | 94 | |
Z93322.1 | - | 3 - 4 | 152 | 94 |