The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004932.3 | NP_004923.1 | 5 - 6 | 1171 | 64 |
XM_011513921.2 | XP_011512223.1 | 5 - 6 | 990 | 64 | |
XM_017008910.1 | XP_016864399.1 | 4 - 5 | 941 | 64 | |
XM_017008911.1 | XP_016864400.1 | 5 - 6 | 1128 | 64 | |
GenBank mRNA | AK291290.1 | - | 5 - 6 | 1137 | 64 |
BC000019.2 | - | 5 - 6 | 1076 | 64 | |
D31784.1 | - | 4 - 5 | 931 | 64 |