Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEPP1
Entrez Gene ID
6414
Gene Name
selenoprotein P, plasma, 1
Gene Aliases
SELP, SEPP, SeP
Chromosome Location
Chr.5: 42799880 - 42811922 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085486.1 NP_001078955.1 5 - 6 660 111
NM_001093726.1 NP_001087195.1 5 - 6 719 111
NM_005410.2 NP_005401.3 4 - 5 631 111
GenBank mRNA AK096125.1 - 4 - 5 1000 111
AK225801.1 - 5 - 6 630 111
AK311392.1 - 4 - 5 968 111
AL833145.1 - 4 - 5 602 111
BC005244.1 - 4 - 5 601 111
BC015875.1 - 4 - 5 605 111
BC040075.1 - 4 - 5 581 111
BC046152.1 - 4 - 5 582 111
BC058919.1 - 4 - 5 612 111
Z11793.1 - 4 - 5 567 111

View Full Product Details