The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243974.1 | NP_001230903.1 | 1 - 2 | 491 | 78 |
NM_001243975.1 | NP_001230904.1 | 1 - 2 | 491 | 78 | |
NM_005605.4 | NP_005596.2 | 1 - 2 | 491 | 78 | |
XM_017013611.1 | XP_016869100.1 | 1 - 2 | 491 | 78 | |
GenBank mRNA | AK055013.1 | - | 1 - 2 | 642 | 78 |
AK299415.1 | - | 1 - 2 | 325 | 78 | |
AY007249.1 | - | 1 - 2 | 98 | 78 | |
BC004864.2 | - | 1 - 2 | 378 | 78 | |
S46622.1 | - | 1 - 2 | 337 | 78 |