Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBXN8
Entrez Gene ID
7993
Gene Name
UBX domain protein 8
Gene Aliases
D8S2298E, REP8, UBXD6
Chromosome Location
Chr.8: 30729131 - 30767006 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282199.1 NP_001269128.1 1 - 2 110 102
NM_005671.3 NP_005662.1 1 - 2 110 102
GenBank mRNA BC020694.1 - 1 - 2 94 102
BI462115.1 - 1 - 2 115 102
BI755534.1 - 1 - 2 86 102
D83767.1 - 1 - 2 100 102

View Full Product Details