The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316374.1 | NP_001303303.1 | 1 - 2 | 445 | 64 |
NM_005917.3 | NP_005908.1 | 1 - 2 | 445 | 64 | |
GenBank mRNA | AK307273.1 | - | 1 - 2 | 56 | 64 |
AK312331.1 | - | 1 - 2 | 86 | 64 | |
BC001484.2 | - | 1 - 2 | 66 | 64 | |
CB266998.1 | - | 1 - 2 | 65 | ||
D55654.1 | - | 1 - 2 | 65 | 64 | |
EU794635.1 | - | 1 - 2 | 66 | 64 |