The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005981.3 | NP_005972.1 | 1 - 2 | 218 | 69 |
GenBank mRNA | AK315765.1 | - | 1 - 2 | 339 | 69 |
AU098749.1 | - | 1 - 2 | 263 | 69 | |
BC010377.1 | - | 1 - 2 | 171 | 69 | |
BX647402.1 | - | 1 - 2 | 196 | 69 | |
CR407647.1 | - | 1 - 2 | 64 | 69 | |
U01160.1 | - | 1 - 2 | 166 | 69 |